siRNA Details
| Gene Symbol: Mgrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013964.1 | NP_001013986.1 | 11, 12 | 1197 | ||
| XM_006245794.2 | XP_006245856.1 | 11, 12 | 1200 | |||
| XM_006245795.2 | XP_006245857.1 | 11, 12 | 1197 | |||
| XM_008767486.1 | XP_008765708.1 | 11, 12 | 1197 | |||
| GenBank mRNA | BC083621.1 | - | 11, 12 | 1197 | 7134018 | |
| HE574489.1 | - | 11, 12 | 1061 | |||

Mgrn1
Genomic Map