siRNA Details
| Gene Symbol: Slx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245835.2 | XP_006245897.2 | 7, 8 | 1740 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slx4| Gene Symbol: Slx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245835.2 | XP_006245897.2 | 7, 8 | 1740 | ||