siRNA Details
| Gene Symbol: Zscan10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106981.1 | NP_001100451.1 | 6 | 1781 | ||
| XM_006245870.2 | XP_006245932.1 | 6 | 1781 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zscan10| Gene Symbol: Zscan10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106981.1 | NP_001100451.1 | 6 | 1781 | ||
| XM_006245870.2 | XP_006245932.1 | 6 | 1781 | |||