siRNA Details
| Gene Symbol: Tnfrsf12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181086.3 | NP_851600.1 | 3 | 350 | ||
| GenBank mRNA | AY255102.1 | - | 3 | 321 | ||
| BC060537.1 | - | 3 | 354 | 6887545 | ||
| FQ229590.1 | - | 3 | 350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfrsf12a| Gene Symbol: Tnfrsf12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181086.3 | NP_851600.1 | 3 | 350 | ||
| GenBank mRNA | AY255102.1 | - | 3 | 321 | ||
| BC060537.1 | - | 3 | 354 | 6887545 | ||
| FQ229590.1 | - | 3 | 350 | |||