siRNA Details
| Gene Symbol: Amdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024990.2 | NP_001020161.2 | 2, 3 | 287 | ||
| XM_008767565.1 | XP_008765787.1 | 2, 3 | 378 | |||
| GenBank mRNA | BC091278.1 | - | 2, 3 | 277 | ||
| CO388545.1 | - | 2, 3 | 287 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172935, BC037005, AK077584, AK090009, AK168432, and others.

Amdhd2
Genomic Map