siRNA Details
| Gene Symbol: Fahd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024991.1 | NP_001020162.1 | 1 | 510 | ||
| GenBank mRNA | BC078950.1 | - | 1 | 510 | 7110058 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fahd1| Gene Symbol: Fahd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024991.1 | NP_001020162.1 | 1 | 510 | ||
| GenBank mRNA | BC078950.1 | - | 1 | 510 | 7110058 | |