siRNA Details
| Gene Symbol: Eme2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750789.3 | XP_003750837.1 | 8 | 1134 | ||
| XM_008767602.1 | XP_008765824.1 | 8 | 1134 | |||
| XM_008767603.1 | XP_008765825.1 | 7 | 1250 | |||
| XM_008767604.1 | XP_008765826.1 | 6 | 1143 | |||
| XM_008767605.1 | XP_008765827.1 | 8 | 1100 | |||
| GenBank mRNA | BC158841.1 | - | 7 | 1231 | ||

Eme2
Genomic Map