siRNA Details
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246057.2 | XP_006246119.1 | 3 | 388 | ||
| XM_008767609.1 | XP_008765831.1 | 3 | 388 | |||
| XM_008767610.1 | XP_008765832.1 | 3 | 298 | |||
| XM_008767611.1 | XP_008765833.1 | 3 | 239 | |||
| GenBank mRNA | FQ211597.1 | - | 3 | 355 | ||

Unkl
Genomic Map