siRNA Details
Gene Symbol: Unkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246057.2 | XP_006246119.1 | 6, 7 | 901 | ||
XM_008767609.1 | XP_008765831.1 | 6, 7 | 901 | |||
XM_008767610.1 | XP_008765832.1 | 6, 7 | 811 | |||
XM_008767611.1 | XP_008765833.1 | 6, 7 | 752 | |||
XM_008767612.1 | XP_008765834.1 | 2, 3 | 641 | |||
GenBank mRNA | FQ211597.1 | - | 6, 7 | 868 |

More Information
Other Species Targeted

Gene Ontology Categories:
