siRNA Details
Gene Symbol: Unkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246057.2 | XP_006246119.1 | 8 | 1022 | ||
XM_008767609.1 | XP_008765831.1 | 8 | 1022 | |||
XM_008767610.1 | XP_008765832.1 | 8 | 932 | |||
XM_008767611.1 | XP_008765833.1 | 8 | 873 | |||
XM_008767612.1 | XP_008765834.1 | 4 | 762 | |||
GenBank mRNA | FQ211597.1 | - | 8 | 989 |
