siRNA Details
| Gene Symbol: Haghl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750790.3 | XP_003750838.1 | 3 | 360 | ||
| XM_006246064.2 | XP_006246126.1 | 3 | 360 | |||
| XM_006246065.1 | XP_006246127.1 | 1 | 35 | |||
| XM_008767617.1 | XP_008765839.1 | 1 | 368 | |||
| GenBank mRNA | BC076383.1 | - | 2 | 256 | ||
| BC161824.1 | - | 2 | 260 | |||

Haghl
Genomic Map