siRNA Details
| Gene Symbol: Capn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106990.1 | NP_001100460.1 | 2 | 1062 | ||
| XM_008767570.1 | XP_008765792.1 | 2 | 8481 | |||
| XM_008767571.1 | XP_008765793.1 | 3 | 1595 | |||
| XM_008767572.1 | XP_008765794.1 | 2 | 8481 | |||
| XM_008767573.1 | XP_008765795.1 | 2 | 8481 | |||
| XM_008767574.1 | XP_008765796.1 | 2 | 8481 | |||
| XM_008767575.1 | XP_008765797.1 | 2 | 8481 | |||
| XM_008767576.1 | XP_008765798.1 | 2 | 1305 | |||

Capn15
Genomic Map