siRNA Details
| Gene Symbol: Tmem8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106991.1 | NP_001100461.1 | 6 | 1059 | ||
| XM_006245973.2 | XP_006246035.1 | 6 | 1673 | |||
| XM_008767577.1 | XP_008765799.1 | 6 | 2751 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem8a| Gene Symbol: Tmem8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106991.1 | NP_001100461.1 | 6 | 1059 | ||
| XM_006245973.2 | XP_006246035.1 | 6 | 1673 | |||
| XM_008767577.1 | XP_008765799.1 | 6 | 2751 | |||