siRNA Details
| Gene Symbol: Crebrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277157.1 | NP_001264086.1 | 3, 4 | 380 | ||
| XM_008767581.1 | XP_008765803.1 | 2, 3 | 600 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crebrf| Gene Symbol: Crebrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277157.1 | NP_001264086.1 | 3, 4 | 380 | ||
| XM_008767581.1 | XP_008765803.1 | 2, 3 | 600 | |||