siRNA Details
| Gene Symbol: Adam19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160228.1 | NP_001153700.1 | 4 | 413 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam19| Gene Symbol: Adam19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160228.1 | NP_001153700.1 | 4 | 413 | ||