siRNA Details
| Gene Symbol: Nipal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106995.1 | NP_001100465.1 | 1, 2 | 121 | ||
| GenBank mRNA | FQ211728.1 | - | 1, 2 | 125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nipal4| Gene Symbol: Nipal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106995.1 | NP_001100465.1 | 1, 2 | 121 | ||
| GenBank mRNA | FQ211728.1 | - | 1, 2 | 125 | ||