siRNA Details
| Gene Symbol: Irgm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012007.1 | NP_001012007.1 | 3 | 1177 | ||
| XM_006246178.2 | XP_006246240.1 | 3 | 1234 | |||
| XM_006246179.2 | XP_006246241.1 | 2 | 1093 | |||
| GenBank mRNA | BC079106.1 | - | 3 | 1177 | 7125255 | |

Irgm
Genomic Map