siRNA Details
Gene Symbol: Maml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106997.1 | NP_001100467.1 | 2 | 1645 | ||
XM_006246251.2 | XP_006246313.1 | 2 | 2919 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Maml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106997.1 | NP_001100467.1 | 2 | 1645 | ||
XM_006246251.2 | XP_006246313.1 | 2 | 2919 |