siRNA Details
| Gene Symbol: Maml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106997.1 | NP_001100467.1 | 2 | 1768 | ||
| XM_006246251.2 | XP_006246313.1 | 2 | 3042 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Maml1| Gene Symbol: Maml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106997.1 | NP_001100467.1 | 2 | 1768 | ||
| XM_006246251.2 | XP_006246313.1 | 2 | 3042 | |||