siRNA Details
| Gene Symbol: Aff4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107001.1 | NP_001100471.1 | 2 | 368 | ||
| XM_006246263.2 | XP_006246325.1 | 2 | 414 | |||
| XM_006246264.2 | XP_006246326.1 | 2 | 337 | |||
| XM_008767684.1 | XP_008765906.1 | 2 | 429 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_033565, BC138999, BC145490, AF190449, AK054401, and others.

Aff4
Genomic Map