siRNA Details
| Gene Symbol: Sept8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107002.1 | NP_001100472.1 | 3 | 290 | ||
| XM_006246265.2 | XP_006246327.1 | 3 | 502 | |||
| XM_006246266.2 | XP_006246328.1 | 3 | 303 | |||
| XM_006246267.2 | XP_006246329.1 | 3 | 303 | |||
| XM_006246268.1 | XP_006246330.1 | 3 | 271 | |||
| XM_006246270.2 | XP_006246332.1 | 3 | 303 | |||
| XM_006246271.2 | XP_006246333.1 | 3 | 303 | |||
| GenBank mRNA | BC158796.1 | - | 3 | 266 | 7314654 | |

Sept8
Genomic Map