siRNA Details
| Gene Symbol: Slc36a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024764.1 | NP_001019935.1 | 5 | 559 | ||
| XM_006246381.2 | XP_006246443.1 | 5 | 602 | |||
| XM_008767685.1 | XP_008765907.1 | 6 | 539 | |||
| GenBank mRNA | BC097463.1 | - | 5 | 559 | 7457579 | |

Slc36a3
Genomic Map