siRNA Details
| Gene Symbol: Atpaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107006.1 | NP_001100476.1 | 8 | 918 | ||
| GenBank mRNA | FQ213754.1 | - | 8 | 909 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atpaf2| Gene Symbol: Atpaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107006.1 | NP_001100476.1 | 8 | 918 | ||
| GenBank mRNA | FQ213754.1 | - | 8 | 909 | ||