siRNA Details
| Gene Symbol: LOC103689956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767884.1 | XP_008766106.1 | 2 | 286 | ||
| Gene Symbol: Map2k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100674.1 | NP_001094144.1 | 2 | 232 | ||
| GenBank mRNA | BC160839.1 | - | 2 | 102 | 7458293 | |
| DY311082.1 | - | 2 | 232 | |||

LOC103689956
Genomic Map