siRNA Details
Gene Symbol: LOC103689956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767884.1 | XP_008766106.1 | 12 | 1193 |
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100674.1 | NP_001094144.1 | 12 | 1139 | ||
GenBank mRNA | BC100095.1 | - | 4 | 258 | ||
BC160839.1 | - | 12 | 1009 | 7458293 | ||
FQ231052.1 | - | 6 | 469 | |||
FQ235176.1 | - | 3 | 81 |
