siRNA Details
| Gene Symbol: Zfp287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107008.2 | NP_001100478.1 | 5 | 1477 | ||
| XM_006246692.2 | XP_006246754.1 | 6 | 1897 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp287| Gene Symbol: Zfp287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107008.2 | NP_001100478.1 | 5 | 1477 | ||
| XM_006246692.2 | XP_006246754.1 | 6 | 1897 | |||