siRNA Details
| Gene Symbol: Trim16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135033.2 | NP_001128505.1 | 5 | 1119 | ||
| XM_008767824.1 | XP_008766046.1 | 5 | 1377 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim16| Gene Symbol: Trim16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135033.2 | NP_001128505.1 | 5 | 1119 | ||
| XM_008767824.1 | XP_008766046.1 | 5 | 1377 | |||