siRNA Details
| Gene Symbol: Cfap52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100968.2 | NP_001094438.2 | 8 | 1003 | ||
| GenBank mRNA | BC161799.1 | - | 8 | 1003 | 7100780 | |
| DQ445463.1 | - | 8 | 1015 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap52| Gene Symbol: Cfap52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100968.2 | NP_001094438.2 | 8 | 1003 | ||
| GenBank mRNA | BC161799.1 | - | 8 | 1003 | 7100780 | |
| DQ445463.1 | - | 8 | 1015 | |||