siRNA Details
| Gene Symbol: Odf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007670.1 | NP_001007671.1 | 2 | 355 | ||
| XM_006246698.2 | XP_006246760.1 | 2 | 777 | |||
| GenBank mRNA | BC079319.1 | - | 2 | 355 | 7135137 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Odf4| Gene Symbol: Odf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007670.1 | NP_001007671.1 | 2 | 355 | ||
| XM_006246698.2 | XP_006246760.1 | 2 | 777 | |||
| GenBank mRNA | BC079319.1 | - | 2 | 355 | 7135137 | |