siRNA Details
| Gene Symbol: TFEB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167827.2 | NP_001161299.2 | 7 | 877 | ||
| NM_001271943.1 | NP_001258872.1 | 6 | 686 | |||
| NM_001271944.1 | NP_001258873.1 | 7 | 941 | |||
| NM_001271945.1 | NP_001258874.1 | 7 | 852 | |||
| NM_007162.2 | NP_009093.1 | 8 | 1043 | |||
| XM_005249411.1 | XP_005249468.1 | 7 | 976 | |||
| XM_005249412.1 | XP_005249469.1 | 7 | 868 | |||
| XM_006715212.3 | XP_006715275.1 | 7 | 1095 | |||
| XM_006715213.2 | XP_006715276.1 | 7 | 919 | |||
| XM_011514915.1 | XP_011513217.1 | 7 | 1098 | |||
| XM_011514916.2 | XP_011513218.1 | 8 | 888 | |||
| GenBank mRNA | AJ608786.1 | - | 7 | 877 | ||
| AK095061.1 | - | 7 | 1968 | |||
| AK298390.1 | - | 7 | 862 | |||
| BC006225.2 | - | 5 | 315 | |||
| BC032448.1 | - | 8 | 1043 | 5180066 | ||
| CT000098.1 | - | 6 | 670 | |||
| M33782.1 | - | 7 | 1023 | |||

TFEB
Genomic Map