siRNA Details
| Gene Symbol: IDH3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004135.3 | NP_004126.1 | 6 | 581 | ||
| NM_174869.2 | NP_777358.1 | 6 | 581 | |||
| GenBank mRNA | AB209206.1 | - | 1 | 3192 | ||
| BC000933.2 | - | 6 | 427 | 3447003 | ||
| BC001902.1 | - | 6 | 422 | 3543839 | ||
| BI831096.1 | - | 6 | 579 | |||
| U40272.1 | - | 6 | 576 | |||
| U69268.1 | - | 2 | 132 | |||
| Z68907.1 | - | 6 | 463 | |||
Citations (1)
Thrombin-cleaved COOH(-) terminal osteopontin peptide binds with cyclophilin C to CD147 in murine breast cancer cells.
- Authors
- Mi Z, Oliver T, Guo H, Gao C, Kuo PC
- Journal
- Cancer Res - Page No 4088 (Tue May 01 00:00:00 UTC 2007)
- Volume (Year)
- 67 (2007)
- Product Usage
- -

IDH3G
Genomic Map