siRNA Details
| Gene Symbol: Pipox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012009.1 | NP_001012009.1 | 3, 4 | 491 | ||
| XM_006246958.1 | XP_006247020.1 | 4, 5 | 627 | |||
| XM_006246959.1 | XP_006247021.1 | 3, 4 | 815 | |||
| GenBank mRNA | BC088249.1 | - | 3, 4 | 491 | 7370364 | |

Pipox
Genomic Map