siRNA Details
Gene Symbol: RGD1307929 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768148.1 | XP_008766370.1 | 6, 7 | 710 | ||
XM_220636.7 | XP_220636.3 | 6, 7 | 710 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1307929 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768148.1 | XP_008766370.1 | 6, 7 | 710 | ||
XM_220636.7 | XP_220636.3 | 6, 7 | 710 |