siRNA Details
| Gene Symbol: Traf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107017.2 | NP_001100487.1 | 7 | 1414 | ||
| GenBank mRNA | BC162047.1 | - | 7 | 1414 | 7107394 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Traf4| Gene Symbol: Traf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107017.2 | NP_001100487.1 | 7 | 1414 | ||
| GenBank mRNA | BC162047.1 | - | 7 | 1414 | 7107394 | |