siRNA Details
| Gene Symbol: Fbxo39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039018.1 | NP_001034107.1 | 2 | 1128 | ||
| XM_006246727.2 | XP_006246789.1 | 3 | 1385 | |||
| XM_006246729.2 | XP_006246791.1 | 3 | 1225 | |||
| GenBank mRNA | BC082089.1 | - | 3 | 1178 | ||
| BC100079.1 | - | 2 | 1128 | 7132945 | ||

Fbxo39
Genomic Map