siRNA Details
| Gene Symbol: Ankfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107018.2 | NP_001100488.2 | 9, 10 | 1193 | ||
| XM_008767830.1 | XP_008766052.1 | 6, 7 | 1541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankfy1| Gene Symbol: Ankfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107018.2 | NP_001100488.2 | 9, 10 | 1193 | ||
| XM_008767830.1 | XP_008766052.1 | 6, 7 | 1541 | |||