siRNA Details
| Gene Symbol: Wsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025664.1 | NP_001020835.2 | 1 | 508 | ||
| NM_001042561.1 | NP_001036026.1 | 6 | 920 | |||
| XM_008768016.1 | XP_008766238.1 | 6 | 1843 | |||
| GenBank mRNA | BC098720.1 | - | 1 | 508 | ||
| BC167746.1 | - | 6 | 923 | 8364583 | ||
| CK473503.1 | - | 4 | 403 | |||

Wsb1
Genomic Map