siRNA Details
| Gene Symbol: Wsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025664.1 | NP_001020835.2 | 3 | 761 | ||
| NM_001042561.1 | NP_001036026.1 | 8 | 1173 | |||
| XM_008768016.1 | XP_008766238.1 | 8 | 2096 | |||
| GenBank mRNA | BC098720.1 | - | 3 | 761 | ||
| BC167746.1 | - | 8 | 1176 | 8364583 | ||

Wsb1
Genomic Map