siRNA Details
| Gene Symbol: Rab11fip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107023.1 | NP_001100493.1 | 8 | 1052 | ||
| XM_006246903.2 | XP_006246965.1 | 8 | 1035 | |||
| XM_006246904.2 | XP_006246966.1 | 6 | 997 | |||
| XM_008768017.1 | XP_008766239.1 | 8 | 929 | |||

Rab11fip4
Genomic Map