siRNA Details
| Gene Symbol: Nsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037189.1 | NP_001032266.1 | 7 | 1189 | ||
| XM_008768018.1 | XP_008766240.1 | 6 | 2337 | |||
| GenBank mRNA | BC099120.1 | - | 7 | 1189 | 7462721 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nsrp1| Gene Symbol: Nsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037189.1 | NP_001032266.1 | 7 | 1189 | ||
| XM_008768018.1 | XP_008766240.1 | 6 | 2337 | |||
| GenBank mRNA | BC099120.1 | - | 7 | 1189 | 7462721 | |