siRNA Details
| Gene Symbol: Psmd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107027.1 | NP_001100497.1 | 8 | 1268 | ||
| XM_006246987.2 | XP_006247049.1 | 8 | 1258 | |||
| XM_006246988.2 | XP_006247050.1 | 8 | 1258 | |||
| GenBank mRNA | BC168749.1 | - | 7 | 737 | ||
| FQ225888.1 | - | 7 | 759 | |||
| FQ230964.1 | - | 3 | 315 | |||

Psmd11
Genomic Map