siRNA Details
| Gene Symbol: Nle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127534.1 | NP_001121006.1 | 7 | 812 | ||
| XM_006246994.2 | XP_006247056.1 | 8 | 1652 | |||
| XM_006246995.2 | XP_006247057.1 | 8 | 1522 | |||
| GenBank mRNA | BC166567.1 | - | 7 | 812 | 7317666 | |

Nle1
Genomic Map