siRNA Details
| Gene Symbol: Slfn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768150.1 | XP_008766372.1 | 5 | 2582 | ||
| XM_220775.6 | XP_220775.3 | 6 | 2449 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slfn5| Gene Symbol: Slfn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768150.1 | XP_008766372.1 | 5 | 2582 | ||
| XM_220775.6 | XP_220775.3 | 6 | 2449 | |||