siRNA Details
| Gene Symbol: Mmp28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079888.1 | NP_001073357.1 | 5, 6 | 843 | ||
| GenBank mRNA | EF121988.1 | - | 5, 6 | 843 | ||
| EF121989.1 | - | 5, 6 | 843 | |||
| Gene Symbol: LOC103694864 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773811.1 | XP_008772033.1 | 5, 6 | 1034 | ||

Mmp28
Genomic Map