siRNA Details
Gene Symbol: Appbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100969.1 | NP_001094439.1 | 10 | 1379 | ||
GenBank mRNA | BC091285.1 | - | 9 | 953 | ||
BC169030.1 | - | 10 | 1380 | 7446141 | ||
EF535262.1 | - | 10 | 1217 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025825, BC019152, AK133853, BC018442, AK214796, and others.
Gene Ontology Categories:
