siRNA Details
| Gene Symbol: TFPI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271003.1 | NP_001257932.1 | 4 | 805 | ||
| NM_006528.3 | NP_006519.1 | 4 | 838 | |||
| GenBank mRNA | AK024817.1 | - | 4 | 600 | ||
| AK075427.1 | - | 4 | 650 | |||
| AK092499.1 | - | 4 | 567 | |||
| AK129833.1 | - | 4 | 600 | |||
| AK313260.1 | - | 4 | 600 | |||
| AY691946.1 | - | 4 | 576 | |||
| BC005330.1 | - | 4 | 600 | 3929933 | ||
| BC034155.1 | - | 3 | 323 | |||
| D29992.1 | - | 4 | 581 | |||
| L27624.1 | - | 4 | 563 | |||

TFPI2
Genomic Map