siRNA Details
| Gene Symbol: Mmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007673.1 | NP_001007674.1 | 7 | 768 | ||
| XM_006247126.1 | XP_006247188.1 | 7 | 806 | |||
| XM_006247127.2 | XP_006247189.1 | 7 | 1628 | |||
| GenBank mRNA | AF540876.1 | - | 7 | 667 | ||
| BC081965.1 | - | 7 | 768 | 7125235 | ||

Mmd
Genomic Map