siRNA Details
| Gene Symbol: Stxbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107038.1 | NP_001100508.1 | 4 | 328 | ||
| XM_006247128.2 | XP_006247190.1 | 3 | 337 | |||
| XM_006247129.2 | XP_006247191.1 | 3 | 320 | |||
| XM_006247130.1 | XP_006247192.1 | 3 | 337 | |||
| XM_006247131.2 | XP_006247193.1 | 3 | 337 | |||
| XM_008768042.1 | XP_008766264.1 | 4 | 429 | |||
| XM_008768043.1 | XP_008766265.1 | 3 | 337 | |||

Stxbp4
Genomic Map