siRNA Details
| Gene Symbol: Utp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135039.1 | NP_001128511.1 | 7 | 967 | ||
| XM_008768044.1 | XP_008766266.1 | 7 | 1222 | |||
| GenBank mRNA | EV776492.1 | - | 5 | 509 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Utp18| Gene Symbol: Utp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135039.1 | NP_001128511.1 | 7 | 967 | ||
| XM_008768044.1 | XP_008766266.1 | 7 | 1222 | |||
| GenBank mRNA | EV776492.1 | - | 5 | 509 | ||