siRNA Details
| Gene Symbol: Dlx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107040.1 | NP_001100510.1 | 1 | 143 | ||
| GenBank mRNA | HQ616894.1 | - | 1 | 165 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlx4| Gene Symbol: Dlx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107040.1 | NP_001100510.1 | 1 | 143 | ||
| GenBank mRNA | HQ616894.1 | - | 1 | 165 | ||